Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, 2.00 M (2.00 N), Ricca Chemical
SDP

APHA for Zinc Testing, EPA for Fluoride Testing

$115.60 - $397.00

Specifications

Chemical Name or Material Sodium Acetate
Grade Laboratory
pH 7.5 to 9.2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
7130-16
SDP
View Documents
Ricca Chemical Company
713016
500 mL Natural Poly Bottle
Each for $115.60
Add to Cart
 
7130-32
SDP
View Documents
Ricca Chemical Company
713032
1 L Natural Poly Bottle
Each for $165.60
Add to Cart
 
R713000010F
SDP
View Documents
Ricca Chemical Company
71302.5
10 L Cubitainer
Each for $397.00
Add to Cart
 
Description

Description

Specifications

Specifications

Sodium Acetate
Laboratory
7.5 to 9.2
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.