Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, 2.00 M (2.00 N), Ricca Chemical
SDP

Catalog No. R713000010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R713000010F 10 L Cubitainer
7130-16 500 mL Natural Poly Bottle
7130-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R713000010F Supplier Ricca Chemical Company Supplier No. 71302.5
Add to Cart
Add to Cart

APHA for Zinc Testing, EPA for Fluoride Testing

Specifications

Chemical Name or Material Sodium Acetate
Name Note 2 Molar/2 Normal
CAS 6131-90-4, 7732-18-5
Assay Percent Range 2.00 ± 0.06 M
Molecular Formula C2H9NaO5
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
Solubility Information Solubility in Water: Infinite
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES CC(=O)[O-].O.O.O.[Na+]
IUPAC Name sodium;acetate;trihydrate
Molecular Weight (g/mol) 136.079
PubChem CID 23665404
ChEBI CHEBI:32138
Grade Laboratory
Odor Odorless to slight acetic acid odor
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH 7.5 to 9.2
Quantity 10 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.