Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, 20% (w/w), Ricca Chemical
SDP

Catalog No. 71204 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71204 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71204 Supplier Ricca Chemical Company Supplier No. 71204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APHA for Iron Testing

Specifications

CAS 6131-90-4, 7732-18-5
CAS Min % 78.0, 19.61
CAS Max % 82.0, 20.41
Molecular Formula C2H9NaO5
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES CC(=O)[O-].O.O.O.[Na+]
IUPAC Name sodium;acetate;trihydrate
Molecular Weight (g/mol) 136.079
PubChem CID 23665404
ChEBI CHEBI:32138
Percent Purity 20%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 7.5 to 9.2
Quantity 120 mL
Concentration or Composition Notes 20% (w/w)
Chemical Name or Material Sodium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.