Learn More
Growcells SODIUM ACETATE 3M 1L EA
Catalog No. 501034811
Encompass_Preferred
Ready-to-use sterile solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered, pH adjusted to 5.5 with Glacial Acetic acid.
Catalog No. 50-103-4811 Supplier Growcells Supplier No. MRGE5265
Ready-to-use sterile solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered, pH adjusted to 5.5 with Glacial Acetic acid.