Learn More
Alkali Scientific Sodium Acetate, Anhydrous, 5 Kg
SDP
Catalog No. 5000661085
Encompass_Preferred
Sodium Acetate, Anhydrous, 5kg, is an essential chemical reagent for laboratory buffer preparation and pH control in biochemical and molecular biology experiments. The 5kg bulk pack is ideal for high-volume applications where consistent pH maintenance is critical. Sodium acetate is widely used in protein separation techniques such as ion exchange chromatography and in various enzymatic assays that require a stable acidic environment. This reagent also acts as a buffering agent in industrial applications like food preservation and chemical synthesis. Its high purity and anhydrous form make it suitable for sensitive experiments in academic and industrial research labs.
Catalog No. 50-006-61085 Supplier Alkali Scientific Supplier No. SOD003CP
Sodium Acetate, Anhydrous, 5kg, is an essential chemical reagent for laboratory buffer preparation and pH control in biochemical and molecular biology experiments. The 5kg bulk pack is ideal for high-volume applications where consistent pH maintenance is critical. Sodium acetate is widely used in protein separation techniques such as ion exchange chromatography and in various enzymatic assays that require a stable acidic environment. This reagent also acts as a buffering agent in industrial applications like food preservation and chemical synthesis. Its high purity and anhydrous form make it suitable for sensitive experiments in academic and industrial research labs.