Learn More
Alkali Scientific Sodium Acetate, Anhydrous, 500 G
SDP
Catalog No. 5000661006
Encompass_Preferred
Sodium Acetate, Anhydrous, is a widely used chemical reagent in laboratory settings, particularly for buffer preparation. This 500g pack is ideal for smaller-scale applications requiring a controlled pH environment. Sodium acetate is often used in biochemical processes such as protein purification, enzymatic reactions, and chromatographic methods. It provides an excellent buffer for maintaining optimal pH conditions in DNA and RNA research. Additionally, it is frequently used in food chemistry as a preservative and flavor agent. Its anhydrous form ensures precise concentrations and minimizes the risk of unwanted moisture interference in laboratory experiments.
Catalog No. 50-006-61006 Supplier Alkali Scientific Supplier No. SOD002CP
Sodium Acetate, Anhydrous, is a widely used chemical reagent in laboratory settings, particularly for buffer preparation. This 500g pack is ideal for smaller-scale applications requiring a controlled pH environment. Sodium acetate is often used in biochemical processes such as protein purification, enzymatic reactions, and chromatographic methods. It provides an excellent buffer for maintaining optimal pH conditions in DNA and RNA research. Additionally, it is frequently used in food chemistry as a preservative and flavor agent. Its anhydrous form ensures precise concentrations and minimizes the risk of unwanted moisture interference in laboratory experiments.