Learn More
Crescent Chemical Co Inc Sodium Acetate Buffer, pH 5.2, 127-09-3, MFCD00012459, 250mL
SDP
Catalog No. 50248395
Encompass_Preferred
Molecular Formula C2H3O2.Na, Molecular Weight 82.034, 3M Solution, Molecular Biology Grade, DNase/RNase not detected. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-395 Supplier Crescent Chemical Co Inc Supplier No. SE39572.01
Molecular Formula C2H3O2.Na, Molecular Weight 82.034, 3M Solution, Molecular Biology Grade, DNase/RNase not detected. Storage Conditions: +15 °C TO +30 °C