Learn More
Aqua Solutions Sodium Acetate Buffer Solution | 10L
SDP
Catalog No. NC1733039
Encompass_Preferred
Composition: 16.472% Acetic Acid (64-19-7), 14.322% Sodium Hydroxide (1310-73-2), 69.205% Water (7732-18-5)
Catalog No. NC1733039 Supplier Aqua Solutions Supplier No. OLI059
Composition: 16.472% Acetic Acid (64-19-7), 14.322% Sodium Hydroxide (1310-73-2), 69.205% Water (7732-18-5)