Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate, MP Biomedicals™

Catalog No. 0219401283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-01283 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-194-01283 Supplier MP Biomedicals, Inc Supplier No. 0219401283
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Acetate is used as a buffer in the buffering range of pH 3.6 to 5.6.

Other reported uses:

  • Used in the purification and precipitation of nucleic acids
  • Used in protein crystallization
  • Staining of gels in protein gel electrophoresis and HPLC
  • Mordant dyeing
  • DNA microarray studies of E. coli response
  • Investigation of protein unfolding during reverse phase chromatography
  • Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents
  • As a substrate of acetokinase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Percent Purity ≥98%
Grade Molecular Biology
Boiling Point >400°C (Decomposition)
Melting Point 324°C (Decomposition)
Molecular Weight (g/mol) 82.03

Specifications

CAS 127-09-3
pH 7.0 to 9.2 (5% aq. soln.)
Quantity 250 g
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Flash Point >250°C (>482°F)
Synonym Acetic Acid Sodium Salt, Anhydrous Sodium Acetate
Solubility Information Soluble freely in water (100 mg/mL). Moderately soluble in alcohol.
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Vapor Pressure 0 hPa at 20°C
Density 1.45 g/cm3
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.