Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium acetate trihydrate, 6131-90-4, MFCD00071557, 250 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S8625250G
Encompass
Linear Formula: CH3COONa · 3H2O, Molecular Weight: 136.08, ReagentPlus, ≥99.0%, Synonym: Acetic acid sodium salt trihydrate.
Catalog No. NC0813536