Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, EMSURE™, MilliporeSigma™

Catalog No. MSX02555 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Plastic Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX02555 12 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX02555 Supplier MilliporeSigma Supplier No. SX02555
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
pH 7.5-9.0
Quantity 12 kg
Flash Point >250°C (anhydrous substance)
Packaging Plastic Drum
Solubility Information 613 g/L
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Grade ACS
Chemical Name or Material Sodium Acetate, Trihydrate
EINECSNumber 204-823-8
RTECSNumber AJ4580000
storageNote1 Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.