Learn More
Medchemexpress LLC Sodium acetate trihydrate | 6131-90-4 | MFCD00071557 | 25g
SDP
Catalog No. 5000266522
Encompass_Preferred
Sodium acetate trihydrate 99 5% is a phase change energy storage material which has characteristics such as appropriate phase change temperature and relatively high latent heat Sodium acetate trihydrate 99 5% can be used in research on heat storage[1][2]
Catalog No. 50-002-66522 Supplier Medchemexpress LLC Supplier No. HYY1325I25G
Sodium acetate trihydrate 99 5% is a phase change energy storage material which has characteristics such as appropriate phase change temperature and relatively high latent heat Sodium acetate trihydrate 99 5% can be used in research on heat storage[1][2]