Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS0250500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Natural Poly Bottle
Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS0250500 500 g Poly Wide Mouth Bottle
RDCS025025A 2.5 kg Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCS0250500 Supplier Ricca Chemical Company Supplier No. RDCS0250500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138

Specifications

Color Colorless to White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals
Quantity 500 g
SMILES CC(=O)[O-].O.O.O.[Na+]
IUPAC Name sodium;acetate;trihydrate
Molecular Weight (g/mol) 136.079
Percent Purity 99 to 101%
Grade ACS
Packaging Poly Wide Mouth Bottle
Chemical Name or Material Sodium Acetate, Trihydrate
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.