Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate for analysis EMSURE™ ACS,ISO,Reag. Ph Eur, MilliporeSigma™
GSA_VA

Catalog No. M1062670500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1062670500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1062670500 Supplier MilliporeSigma Supplier No. 1.06267.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent suitable for analytical applications and use as an enzyme substrate.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
Physical Form Crystalline Solid
Quantity 500 g
Ignition Point 607°C
Packaging Plastic Bottle
Density Bulk Density: 900 kg/m3
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Grade ACS/ISO/Reagent Ph. Eur.
Chemical Name or Material Sodium acetate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.