Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1062679050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1062679050 50 kg Plastic Bottle
M1062671000 1 kg Plastic Bottle
M1062675000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1062679050 Supplier MilliporeSigma Supplier No. 1062679050
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
Color Colorless
pH 7.5 to 9.2 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 50 kg
Solubility Information 613g/L
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Grade ACS
Packaging Plastic Bottle
Density 1.42 g/3 (20 °C)
Chemical Name or Material Sodium Acetate Trihydrate
Show More Show Less
EINECSNumber 204-823-8
RTECSNumber AJ4580000
Recommended Storage +5°C to +30°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.