Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, MACCO

Catalog No. 502985655
Encompass
Change view
Click to view available options
Quantity:
12 kg
38.6 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5655 50 kg
50-298-5612 12 kg
50-298-5654 38.6 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-298-5655 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M51101030500

Please call customer service for current pricing. 1-800-766-7000

Chemical Identifiers

CAS 6131-90-4
Molecular Weight (g/mol) 136.08 g/mol

Specifications

Physical Form Crystalline Powder
Quantity 50 kg
Percent Purity 99 to 100%
Grade USP
Packaging 50 kg poly drum with 2xPE liner
Chemical Name or Material Sodium acetate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.