Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, MP Biomedicals

Catalog No. MP023001845 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP023001845 500 g
MP230018401 1 kg
MP230018406 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP023001845 Supplier MP Biomedicals, Inc Supplier No. 02300184.5
Add to Cart
Add to Cart

  • To 800-900 mL deionized water add: 8.20 g sodium acetate and mix
  • Adjust to pH 5.2 with 1.0 M NaOH
  • QS to 1 L with deionized water
  • Mix

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 56°C to 64°C (literature)
Color White
Boiling Point 123°C (-3H)
Linear Formula CH3COONa . 3H2O
Quantity 500 g
Solubility Information Soluble in water (20 g plus 150 mL).(Lit.)
Formula Weight 136.08
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material SODIUM ACETATE TRIHYDRATE
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.