Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium acetate trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Catalog No. 60046131 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
6 x 1 kg
6 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-131 6 x 500 g
60-024-23 500 g
60-024-21 1 kg
60-024-22 2.5 kg
60-046-129 6 x 1 kg
60-046-130 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-046-131 Supplier Solstice Supplier No. 323186X500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 6 x 500 g
Linear Formula CH3COONa · 3H2O
MDL Number MFCD00071557
UN Number NONH for all modes of transport
Formula Weight 136.08
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
EINECSNumber 204-823-8
RTECSNumber AJ4580000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.