Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acetate Trihydrate, USP, EP, JP, FCC, Pyrogen Free, Macco Organiques
SDP

Catalog No. 18614842 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Plastic Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-842 12 kg Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-614-842 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. 51101030120KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macco products offered by Spectrum™ Chemical are suitable for biopharmaceutical production.

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

Color Clear/Colorless
pH 7.5 to 9
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Percent Purity 99 to 101%
Grade EP/FCC/JP/USP
Loss on Drying 40.5%
Packaging Plastic Pail
Chemical Name or Material Sodium Acetate Trihydrate, Pyrogen Free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.