Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Acid Pyrophosphate, FCC, 93-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606281 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-281 45 kg Poly Bottle
18-606-278 500 g Poly Bottle
18-606-279 2.5 kg Poly Bottle
18-606-280 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-281 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S109945KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1099, 7758-16-9, Na2H2P2O7

Spectrum™ Chemical Sodium Acid Pyrophosphate, FCC is leavening agent found in baking powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-16-9
Molecular Formula Na4O7P2
Molecular Weight (g/mol) 265.90
InChI Key FQENQNTWSFEDLI-UHFFFAOYSA-J
IUPAC Name tetrasodium (phosphonatooxy)phosphonate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Percent Purity 93 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Acid Pyrophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.