Learn More
Medchemexpress LLC Sodium alginate | 9005-38-3 | 32,000-200,000 g/mol | (C6H7NaO6)n | 10g
SDP
Catalog No. 5000266887
Encompass_Preferred
Sodium alginate is the sodium salt of alginic acid Sodium alginate can be extracted and purified from brown seaweed Laminaria japonica Sodium alginate can be used in food additives and pharmaceuticals adsorb heavy metal ions and has mucosal-protective and hemostatic effects[1][2]
Catalog No. 50-002-66887 Supplier Medchemexpress LLC Supplier No. HYY131010G
Sodium alginate is the sodium salt of alginic acid Sodium alginate can be extracted and purified from brown seaweed Laminaria japonica Sodium alginate can be used in food additives and pharmaceuticals adsorb heavy metal ions and has mucosal-protective and hemostatic effects[1][2]