Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Alkylnaphthalenesulfonate (ca. 40% in Water), TCI America™
SDP

Catalog No. B0730500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0730500G 500 g
B073025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0730500G Supplier TCI America Supplier No. B0730500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym Alkylnaphthalenesulfonic Acid Sodium Salt, Sodium Butylnaphthalenesulfonate (so called), Butylnaphthalenesulfonic Acid Sodium Salt (so called)

Specifications

Color Brown
Quantity 500 g
Physical Form Liquid
Chemical Name or Material Sodium Alkylnaphthalenesulfonate (ca. 40% in Water)
EINECSNumber (4)-0472
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.