Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ammonium Phosphate Tetrahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. S218500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S218-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S218-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7783-13-3
Molecular Formula H13NNaO8P
Molecular Weight (g/mol) 209.067
MDL Number MFCD00066970
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
PubChem CID 56924540
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

Specifications

Melting Point 79°C
Color Colorless
pH 8
Physical Form Solid
Quantity 500 g
Linear Formula NaNH4HPO4·4H2O
Formula Weight 209.07
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.54g/cm³
Chemical Name or Material Sodium Ammonium Phosphate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.