Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Arsenate, Heptahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18613264 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
10 kg
25 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-264 25 kg Poly Bottle
18-613-259 25 g Poly Bottle
18-613-260 100 g Poly Bottle
18-613-261 250 g Poly Bottle
18-613-262 1 kg Poly Bottle
18-613-263 10 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-613-264 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S113025KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10048-95-0

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 25 kg
Molecular Weight (g/mol) 312.01
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Arsenate, Heptahydrate, Crystal
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.