Learn More
Apexbio Technology LLC Sodium ascorbate 134-03-2 10mM (in 1mL H2O)
SDP
Catalog No. 5000063799
Encompass_Preferred
Sodium ascorbate (CAS 134-03-2) is the sodium salt derivative of ascorbic acid (vitamin C) offering improved bioavailability relative to the free acid form Acting primarily as an antioxidant sodium ascorbate donates electrons to neutralize reactive oxygen species thus mitigating oxidative stress within biological systems Moreover it functions as a cofactor for enzymatic reactions involved in collagen synthesis neurotransmitter biosynthesis and regulation of immune function In biomedical research sodium ascorbate is employed to investigate antioxidant defense mechanisms cellular redox status and the therapeutic potential of vitamin C supplementation in various models
Catalog No. 50-000-63799 Supplier Apexbio Technology LLC Supplier No. B18345.1
Sodium ascorbate (CAS 134-03-2) is the sodium salt derivative of ascorbic acid (vitamin C) offering improved bioavailability relative to the free acid form Acting primarily as an antioxidant sodium ascorbate donates electrons to neutralize reactive oxygen species thus mitigating oxidative stress within biological systems Moreover it functions as a cofactor for enzymatic reactions involved in collagen synthesis neurotransmitter biosynthesis and regulation of immune function In biomedical research sodium ascorbate is employed to investigate antioxidant defense mechanisms cellular redox status and the therapeutic potential of vitamin C supplementation in various models