Learn More
U.S. Pharmacopeia Sodium ascorbate, 134-03-2, MFCD00082340, 200 mg
Catalog No. NC1348614
Encompass
Empirical Formula (Hill Notation): C6H7NaO6, Molecular Weight: 198.11, mp: 220 °C, Synonym: (+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt.
Catalog No. NC1348614 Supplier U.S. Pharmacopeia Supplier No. 1613509
Empirical Formula (Hill Notation): C6H7NaO6, Molecular Weight: 198.11, mp: 220 °C, Synonym: (+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt.