Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Ascorbate, FCC, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606304 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-304 50 kg Fiber Drum
18-606-302 500 g Amber Glass Bottle
18-606-303 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-304 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S113750KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1137, 134-03-2, C6H7NaO6

Spectrum™ Chemical Sodium Ascorbate, FCC is used as a food additive as an antioxidant and acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

CAS Max % 1
Loss on Drying 0.0025
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 99 to 101%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.