Learn More
Medchemexpress LLC Sodium aurothiomalate | 12244-57-4 | MFCD00064304 | 98.0% | 371.11 g/mol | C4H6O4S.Au.xNa | 50 MG
SDP
Catalog No. 5000242644
Encompass_Preferred
Aurothiomalate sodium is a gold-containing compound used in research as a selective inhibitor of oncogenic PKCι signaling and as a thioredoxin reductase (TrxR) inhibitor. Historically used as an anti-rheumatic agent, it has reported anti-tumor activity and is applied in biochemical and cell-based studies.
  • Potent inhibitor of PKCι signaling and thioredoxin reductase
  • Shows anti-tumor and anti-inflammatory activity in studies
  • Supplied as a solid, off-white to light yellow material
  • Typical purity around 98.0%
  • Stable under refrigerated, sealed storage away from moisture
  • Available in small milligram quantities for research use
Catalog No. 50-002-42644 Supplier Medchemexpress LLC Supplier No. HY10638150MG
Aurothiomalate sodium is a gold-containing compound used in research as a selective inhibitor of oncogenic PKCι signaling and as a thioredoxin reductase (TrxR) inhibitor. Historically used as an anti-rheumatic agent, it has reported anti-tumor activity and is applied in biochemical and cell-based studies.
  • Potent inhibitor of PKCι signaling and thioredoxin reductase
  • Shows anti-tumor and anti-inflammatory activity in studies
  • Supplied as a solid, off-white to light yellow material
  • Typical purity around 98.0%
  • Stable under refrigerated, sealed storage away from moisture
  • Available in small milligram quantities for research use