Learn More
Medchemexpress LLC Sodium aurothiomalate | 12244-57-4 | MFCD00064304 | 98.0% | 390.08 g/mol | C4H3AuNa2O4S | 10 MG
SDP
Catalog No. 5000243408
Encompass_Preferred
Sodium aurothiomalate is a gold-containing small molecule supplied for research use. It functions as an inhibitor of oncogenic PKCι signaling and thioredoxin reductase, and has been used historically as an anti-rheumatic agent and in antitumor studies. Supplied as an off-white to light-yellow solid with high purity and available in mg-scale packages.
  • Gold-containing thiomalate salt with defined biological activity.
  • Potent inhibitor of PKCι signaling and thioredoxin reductase.
  • High purity suitable for biochemical and cellular studies.
  • Solid form, off-white to light yellow, for straightforward handling.
  • Available in small mg-scale package sizes for research workflows.
Catalog No. 50-002-43408 Supplier Medchemexpress LLC Supplier No. HY10638110MG
Sodium aurothiomalate is a gold-containing small molecule supplied for research use. It functions as an inhibitor of oncogenic PKCι signaling and thioredoxin reductase, and has been used historically as an anti-rheumatic agent and in antitumor studies. Supplied as an off-white to light-yellow solid with high purity and available in mg-scale packages.
  • Gold-containing thiomalate salt with defined biological activity.
  • Potent inhibitor of PKCι signaling and thioredoxin reductase.
  • High purity suitable for biochemical and cellular studies.
  • Solid form, off-white to light yellow, for straightforward handling.
  • Available in small mg-scale package sizes for research workflows.