Learn More
Sigma Aldrich Sodium aurothiomalate hydrate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Sodium aurothiomalate has antirheumatic properties and exhibits inhibitory effects against m
- Tuberculosis.sodium aurothiomalate hydrate was used in the high mobility group box chromosomal protein 1 (hmgb1) western blotting
Specifications
Specifications
| Molecular Formula | C4H3AuNa2O4S -+ xH2O |
| Linear Formula | NaO2CCH2CH(SAu)CO2Na -+ xH2O |
| MDL Number | MFCD00166971 |
| Quantity | 250 mg |
| Synonym | (1,2-Dicarboxyethylthio)gold disodium salt hydrate; Gold sodium thiomalate hydrate; Mercaptobutanedioic acid gold(I) disodium salt |
| Molecular Weight (g/mol) | 390.08 (anhydrous basis) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.