Learn More
MOLTOX Molecular Toxicology Inc Sodium Azide, 100 ug/vial, 5 vials/pack, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60103.3
Encompass
For use in Ames test with TA100 & TA1535 tester strains. Dose 1.5 - 5.0 µg/plate. Reconstitute with sterile dH2O as applicable. Direct acting mutagens do not require the use of S9 metabolic activation to cause mutagenicity. CONTROLCHEM™ chemicals are obtained from major suppliers of research chemicals and are employed without further characterization or purification. Packaged quantities are precise within 1%. Please Note: CONTROLCHEM™ chemicals are known mutagens/carcinogens/toxins and are sold only to those experienced in the safe handling and disposal of hazardous materials. Consult your Institutional Safety Officer before ordering.
Catalog No. 50-948-870