Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Azide, 5% (w/v), Ricca Chemical
SDP

Catalog No. 7144832 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7144-832 1 L Natural Poly Bottle
71448-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7144-832 Supplier Ricca Chemical Company Supplier No. 7144.832
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26628-22-8 , 7732-18-5
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

CAS Min % 93.0, 4.67
CAS Max % 97.0, 4.86
Percent Purity 5%
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
Quantity 1 L
Chemical Name or Material Sodium Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.