Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Azide, ≥99%, (Crystalline), MP Biomedicals

Catalog No. ICN10289190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10289190 500 g
ICN10289125 25 g
ICN10289180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10289190 Supplier MP Biomedicals, Inc Supplier No. 0210289190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26628-22-8 | Formula: N3Na | Weight: 65.011 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Physical Form Crystalline
Quantity 500 g
Assay Percent Range ≥99%
Formula Weight 65
Chemical Name or Material Sodium Azide
storageNote1 Not for export
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.