Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Azide, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613284 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-284 50 kg Fiber Drum
18-613-280 100 g Amber Glass Bottle
18-613-282 2.5 kg Amber Glass Bottle
18-613-281 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-613-284 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S166650KGBL
Only null left
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
IUPAC Name sodium azide
SMILES [Na+].[N-]=[N+]=[N-]

Specifications

CAS Max % 1
Quantity 50 kg
Percent Purity 99%
Grade ACS
Loss on Drying 0.001
Packaging Fiber Drum

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.