Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Azide, MP Biomedicals

Catalog No. MP210289110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210289110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210289110 Supplier MP Biomedicals, Inc Supplier No. 0210289110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Sodium azide used as a preservative for laboratory reagents; a bacteriostatic agent in storage solutions
  • Inhibits gram negative flora so is therefore included in culture media for the selective isolation of Streptococci and Staphylococci
  • It has been used to avoid bacterial contamination during RPC-5 column chromatography of DNA fragments

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Melting Point >300°C (decomposition)
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula NaN3
Solubility Information Soluble in water (100mg/mL - clear, colorless solution); slightly soluble in alcohol and ethanol; insoluble in ether.
Formula Weight 65
Percent Purity ≥99%
Chemical Name or Material SODIUM AZIDE
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.