Learn More
GFS Chemicals Inc SODIUM AZIDE, REAGENT (ACS) | 100 G | GFS CHEMICALS, INC. Item #: 66521 | Bulk #: 3512
SDP
Catalog No. NC0904192
Encompass_Preferred
SODIUM AZIDE, REAGENT (ACS) | Chemical Formula : NaN!3 | CAS # : 26628-22-8 | Specifications : Assay 99.0 % - Min, Insoluble matter 0.05 % , Loss on drying (at 105 C) (%) 0.1 % , Titrable base 0.05 meq/g
Catalog No. NC0904192 Supplier GFS Chemicals Inc Supplier No. 66521
SODIUM AZIDE, REAGENT (ACS) | Chemical Formula : NaN!3 | CAS # : 26628-22-8 | Specifications : Assay 99.0 % - Min, Insoluble matter 0.05 % , Loss on drying (at 105 C) (%) 0.1 % , Titrable base 0.05 meq/g