Learn More
Andwin Scientific SODIUM AZIDE SOLUTION 5% 500ML
SDP
Supplier:  Andwin Scientific BDH74652
Encompass_Preferred
The Sodium Azide Solution is a clear liquid chemical that is classified as hazardous and toxic. This chemical has an assay of 4.90 - 5.10 %. It is a N.I.S.T. traceable solution and is composed of purified water. This is packaged and sold in a 500 mL poly bottle. It has a shelf life of 2 years.
Catalog No. NC0690399