Learn More
Medchemexpress LLC Sodium benzenesulfinate | 873-55-2 | 99.9% | 164.16 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013598500G
Encompass_Preferred
Sodium benzenesulfinate is a reagent used in organic synthesis to create other compounds, such as sulfones. It undergoes rhodium-catalyzed desulfinative coupling with aldehydes to yield ketones. This product is a solid, white to off-white in appearance.
  • Used in organic synthesis
  • Undergoes rhodium-catalyzed desulfinative coupling
  • Available as a solid
  • White to off-white color
  • Stable under recommended storage conditions
  • Incompatible with strong acids/alkalis, strong oxidizing/reducing agents
Catalog No. 50-004-54592