Learn More
Medchemexpress LLC Sodium benzenesulfonate | 515-42-4 | MFCD00065179 | >96.0% | 180.16 g/mol | C6H5NaO3S | 500 G

Supplier: Medchemexpress LLC HYW016510500G
Sodium benzenesulfonate (CAS 515-42-4) is the sodium salt of benzenesulfonic acid used as a biochemical reagent and analytical standard. It is supplied as a white to off-white solid for research and analytical applications requiring a sulfonated aromatic salt.
- Chemical formula C6H5NaO3S, molecular weight 180.16 g/mol.
- White to off-white solid appearance.
- Purity greater than 96.0% (HPLC) for reagent-grade material.
- Available in multiple packaging sizes, including 500 g.
- Commonly used as a reagent and analytical standard in research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.