Learn More
Medchemexpress LLC Sodium benzoate | 532-32-1 | MFCD00012463 | 99.6% | 144.10 g·mol-1 | C7H5NaO2 | 100mg
SDP
Catalog No. 5000266598
Encompass_Preferred
Sodium benzoate is an orally active pharmaceutical excipient such as an antibacterial agent preservative lubricant etc Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs Sodium benzoate activates NF- B and induces Apoptosis Sodium benzoate induces immune suppression and produces reproductively toxic Sodium benzoate can be used for colon cancer and immune disease research[1][2][3][4][5]
Catalog No. 50-002-66598 Supplier Medchemexpress LLC Supplier No. HYY1316100MG
Sodium benzoate is an orally active pharmaceutical excipient such as an antibacterial agent preservative lubricant etc Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs Sodium benzoate activates NF- B and induces Apoptosis Sodium benzoate induces immune suppression and produces reproductively toxic Sodium benzoate can be used for colon cancer and immune disease research[1][2][3][4][5]