Learn More
Selleck Chemical LLC Sodium benzoate S3750-25mg
SDP
Catalog No. 503108373
Encompass_Preferred
Sodium benzoate (Benzoic acid sodium salt) is a preservative and uesd as a food additive which is bacteriostatic and fungistatic under acidic conditions
Catalog No. 50-310-8373 Supplier Selleck Chemical LLC Supplier No. S375025MG
Sodium benzoate (Benzoic acid sodium salt) is a preservative and uesd as a food additive which is bacteriostatic and fungistatic under acidic conditions