Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium beta-Styrenesulfonate 95.0+%, TCI America™
SDP

Catalog No. S02285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02285G Supplier TCI America Supplier No. S02285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2039-44-3
Molecular Formula C8H7NaO3S
Molecular Weight (g/mol) 206.191
MDL Number MFCD00053626
InChI Key MNCGMVDMOKPCSQ-UHDJGPCESA-M
Synonym beta-Styrenesulfonic Acid Sodium Salt
PubChem CID 23678061
IUPAC Name sodium;(E)-2-phenylethenesulfonate
SMILES C1=CC=C(C=C1)C=CS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 206.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium beta-Styrenesulfonate
EINECSNumber (3)-1948
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.