Learn More
American Radiolabeled Chemicals Inc SODIUM BICARB-14C 1LC
SDP
Catalog No. 5090105429
Encompass_Preferred
Sodium bicarbonate-14C 1mCi in sterile water, 1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 84.0
Catalog No. 50-901-05429 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 0138A1MC
Sodium bicarbonate-14C 1mCi in sterile water, 1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 84.0