Learn More
Alkali Scientific Sodium Bicarbonate, 1 Kg
SDP
Supplier:  Alkali Scientific UMB001CP
Encompass_Preferred
Sodium Bicarbonate (1kg, Laboratory Grade) is a versatile chemical reagent widely used in laboratories for neutralization reactions, pH buffering, and as a mild base in various chemical procedures. This 1kg container is ideal for general laboratory use, including preparation of sodium bicarbonate solutions for acid-base titrations. Sodium bicarbonate is also used in pharmaceutical formulations, food testing, and as a mild abrasive in laboratory cleaning. It plays a key role in maintaining stable pH levels in buffers and in applications where a weak alkaline medium is needed for chemical reactions or biological studies.
Catalog No. 50-006-60457