Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, 1 M Buffer Solution, pH 8.5 PROMO

Catalog No. J60408APF25
Change view
Click to view available options
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J60408APF25 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J60408APF25 Supplier Thermo Scientific Chemicals Supplier No. J60408APPR/DEL

missing translation for 'validMainframeMsg'

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 to 7.6. It is also used as a nutrient source.

  • Applications: This solution prepared in water is used as an injection in the pharmaceutical industry and is used to treat cement contamination in shore pac drilling slurry and bentonite water based drilling fluids. It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.
  • Solubility: Soluble in water
  • Notes: Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01 g/mol
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name Sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in Water
Chemical Name or Material Sodium bicarbonate Buffer Solution
TSCA Yes
Recommended Storage Ambient Temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.