Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Isotope Laboratories Sodium Bicarbonate (13C, 99%), 1g

Catalog No. NC0120715
Encompass

Sodium Bicarbonate (13C, 99%), 87081-58-1, NaH*CO3, 85.00, 1g

Catalog No. NC0120715 Supplier Cambridge Isotope Laboratories Supplier No. CLM4411
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Bicarbonate (13C, 99%), 87081-58-1, NaH*CO3, 85.00, 1g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.