Learn More
Medchemexpress LLC Sodium bicarbonate | 144-55-8 | MFCD00003528 | 99.0% | 84.01 g/mol | NaHCO3 | 100 MG
SDP
Catalog No. 5000281789
Encompass_Preferred
Sodium bicarbonate is an inorganic salt used as a buffering agent for cell culture and laboratory applications. It helps maintain physiological pH in culture media and is supplied as a solid for preparation of aqueous stock solutions (CAS 144-55-8; formula NaHCO3). The material is water-soluble and hygroscopic; follow standard handling and storage practices.
  • Cell culture buffer for pH stabilization.
  • Soluble in water for easy stock solution preparation.
  • Useful for antacid and buffering studies in vitro.
  • Supports preparation of precise molar concentrations.
  • Hygroscopic - store in a dry, closed container.
Catalog No. 50-002-81789 Supplier Medchemexpress LLC Supplier No. HYW251598C100MG
Sodium bicarbonate is an inorganic salt used as a buffering agent for cell culture and laboratory applications. It helps maintain physiological pH in culture media and is supplied as a solid for preparation of aqueous stock solutions (CAS 144-55-8; formula NaHCO3). The material is water-soluble and hygroscopic; follow standard handling and storage practices.
  • Cell culture buffer for pH stabilization.
  • Soluble in water for easy stock solution preparation.
  • Useful for antacid and buffering studies in vitro.
  • Supports preparation of precise molar concentrations.
  • Hygroscopic - store in a dry, closed container.