Learn More
Sodium bicarbonate, 1M buffer soln., pH 8.0
Catalog No. AAJ62495AK
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
AAJ62495AK 250 mL
1 options
Catalog No. AAJ62495AK Supplier Thermo Scientific Chemicals Supplier No. J62495AK
Add to Cart
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Liquid
Quantity 250 mL