Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium bicarbonate, 144-55-8, MFCD00003528, 500g
Catalog No. 501656908
Encompass
Linear Formula NaHCO3, Molecular Weight 84.01, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture, Synonym: Sodium hydrogen carbonate
Catalog No. 50-165-6908 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S5761500G
Linear Formula NaHCO3, Molecular Weight 84.01, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture, Synonym: Sodium hydrogen carbonate