Learn More
Hach Company Sodium Bicarbonate, ACS, 454 g
Catalog No. 501994447 Shop All Hach Company Products
Encompass
NaHCO3, ACS Reagent Grade, CAS No. 144-55-8, FW 84.01 Assay 99.7 to 100.3%
Catalog No. 50-199-4447 Supplier Hach Company Supplier No. 77601
NaHCO3, ACS Reagent Grade, CAS No. 144-55-8, FW 84.01 Assay 99.7 to 100.3%